Assay Details
| Gene Symbol: MIR23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038539.1 | 1 | 35 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aminopeptidase O (putative) | AOPEP |
C10H9orf3, C9orf3 |
Sense | Chr. 10 - 27078115 - 27472347 on NCBI Build Sscrofa11.1 | |
| Downstream | fructose-bisphosphatase 1 | FBP1 | FBP | 508.3kb | Anti-sense | Chr. 10 - 27588609 - 27622225 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR23B
Genomic Map