Assay Details
| Gene Symbol: MIR27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038536.1 | 1 | 41 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aminopeptidase O (putative) | AOPEP |
C10H9orf3, C9orf3 |
Sense | Chr. 10 - 27078115 - 27472347 on NCBI Build Sscrofa11.1 | |
| Downstream | microRNA mir-23b | MIR23B | ssc-mir-23b | 155.0 | Sense | Chr. 10 - 27080197 - 27080276 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR27B
Genomic Map