Assay Details
| Gene Symbol: MIR504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038561.1 | 1 | 36 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-9809 | MIR9809 | ssc-mir-9809 | -23.0kb | Sense | Chr. X - 113472239 - 113472331 on NCBI Build Sscrofa11.1 |
| Downstream | 40S ribosomal protein S27-like | LOC100523110 | 575.5kb | Sense | Chr. X - 114070842 - 114071361 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR504
Genomic Map