Assay Details
| Gene Symbol: MIR24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038490.1 | 1 | 33 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-27a | MIR27A | ssc-mir-27a | -78.0 | Sense | Chr. 2 - 65308294 - 65308375 on NCBI Build Sscrofa11.1 |
| Downstream | zinc finger SWIM-type containing 4 | ZSWIM4 | 2.7kb | Anti-sense | Chr. 2 - 65311198 - 65336965 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR24-1
Genomic Map