Assay Details
| Gene Symbol: MIR30E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038525.1 | 1 | 40 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-30c-1 | MIR30C-1 | ssc-mir-30c-1 | -3.0kb | Sense | Chr. 6 - 170426781 - 170426860 on NCBI Build Sscrofa11.1 |
| Downstream | regulating synaptic membrane exocytosis 3 | RIMS3 | 76.5kb | Anti-sense | Chr. 6 - 170506392 - 170546939 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR30E
Genomic Map