Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987588.2 | XP_013843042.2 | 398 | 76 | ||
| XM_021084674.1 | XP_020940333.1 | 517 | 76 | |||
| XM_021084675.1 | XP_020940334.1 | 517 | 76 | |||
| XM_021084676.1 | XP_020940335.1 | 517 | 76 | |||
| XM_021084677.1 | XP_020940336.1 | 373 | 76 | |||
| XM_021084678.1 | XP_020940337.1 | 379 | 76 | |||
More Information
Set Membership: |
|

PRR16
Genomic Map