Assay Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482931.4 | XP_003482979.1 | 470 | 85 | ||
| GenBank mRNA | AK237269.1 | - | 1 - 2 | 429 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482931.4 | XP_003482979.1 | 470 | 85 | ||
| GenBank mRNA | AK237269.1 | - | 1 - 2 | 429 | 85 | |
Set Membership: |
|