Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244747.1 | NP_001231676.1 | 5 - 6 | 493 | 103 | |
| XM_005673419.3 | XP_005673476.1 | 475 | 103 | |||
| XM_005673420.3 | XP_005673477.1 | 487 | 103 | |||
| XM_005673422.3 | XP_005673479.1 | 386 | 103 | |||
| XM_005673423.3 | XP_005673480.1 | 398 | 103 | |||
| XM_005673424.3 | XP_005673481.1 | 295 | 103 | |||
| XM_005673425.3 | XP_005673482.1 | 307 | 103 | |||
| XM_013985906.2 | XP_013841360.1 | 462 | 103 | |||
| XM_021079457.1 | XP_020935116.1 | 1043 | 103 | |||
| XM_021079458.1 | XP_020935117.1 | 464 | 103 | |||
| XM_021079459.1 | XP_020935118.1 | 396 | 103 | |||
| XM_021079461.1 | XP_020935120.1 | 384 | 103 | |||
| GenBank mRNA | AK236335.1 | - | 5 - 6 | 493 | 103 | |
| AK350247.1 | - | 2 - 3 | 242 | 103 | ||
| AK393047.1 | - | 2 - 3 | 260 | 103 | ||
| AK394939.1 | - | 2 - 3 | 248 | 103 | ||
| AY609661.1 | - | 1 - 2 | 493 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244747.1, XM_005673419.3, XM_005673420.3, XM_005673422.3, XM_005673423.3, XM_005673424.3, XM_005673425.3, XM_013985906.2, XM_021079457.1, XM_021079458.1, XM_021079459.1, XM_021079461.1 transcripts.
Set Membership: |
|

RAB9A
Genomic Map