Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244791.1 | NP_001231720.1 | 1 - 2 | 397 | 106 | |
| XM_013980185.2 | XP_013835639.1 | 427 | 106 | |||
| XM_013980186.2 | XP_013835640.1 | 495 | 106 | |||
| GenBank mRNA | AK235751.1 | - | 1 - 2 | 387 | 106 | |
| AK348633.1 | - | 1 - 2 | 397 | 106 | ||
| AK389517.1 | - | 1 - 2 | 387 | 106 | ||
| AK397491.1 | - | 1 - 2 | 457 | 106 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map