Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244928.1 | NP_001231857.1 | 1 - 2 | 83 | 108 | |
| XM_021068422.1 | XP_020924081.1 | 219 | 108 | |||
| GenBank mRNA | AY610369.1 | - | 1 - 2 | 83 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244928.1, XM_021068422.1 transcripts.
Set Membership: |
|

LSM3
Genomic Map