Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256367.1 | NP_001243296.1 | 3 - 4 | 455 | 67 | |
| GenBank mRNA | AY609524.1 | - | 3 - 4 | 356 | 67 | |
| JN606069.1 | - | 3 - 4 | 456 | 67 | ||
| JQ026234.1 | - | 3 - 4 | 455 | 67 | ||
More Information
Set Membership: |
|

NPY
Genomic Map