Assay Details
| Gene Symbol: UBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105309.1 | NP_001098779.1 | 1 - 2 | 86 | 111 | |
| GenBank mRNA | AK232073.1 | - | 1 - 2 | 363 | 111 | |
| AK232547.1 | - | 1 - 2 | 98 | 111 | ||
| AK233993.1 | - | 1 - 2 | 98 | 111 | ||
| AK345386.1 | - | 1 - 2 | 98 | 111 | ||
| AK346579.1 | - | 1 - 2 | 98 | 111 | ||
| AK350322.1 | - | 1 - 2 | 406 | 111 | ||
| AK392357.1 | - | 1 - 2 | 99 | 111 | ||
| AK392400.1 | - | 1 - 2 | 99 | 111 | ||
| AK393787.1 | - | 1 - 2 | 105 | 111 | ||
| AK394386.1 | - | 1 - 2 | 93 | 111 | ||
| AK394470.1 | - | 1 - 2 | 391 | 111 | ||
| AK398053.1 | - | 1 - 2 | 391 | 111 | ||
| AK399318.1 | - | 1 - 2 | 244 | 111 | ||
| AK399439.1 | - | 1 - 2 | 127 | 111 | ||
| EF688558.1 | - | 1 - 2 | 86 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105309.1 transcripts.
Set Membership: |
|

UBB
Genomic Map