Assay Details
| Gene Symbol: DOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143705.2 | NP_001137177.2 | 1 - 2 | 103 | 84 | |
| GenBank mRNA | FJ438468.1 | - | 1 - 2 | 76 | 84 | |
| HX211042.1 | - | 1 - 2 | 104 | 84 | ||
More Information
Set Membership: |
|

DOK1
Genomic Map