Assay Details
| Gene Symbol: MPEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267892.1 | NP_001254821.1 | 1 | 2215 | 69 | |
| GenBank mRNA | AK350421.1 | - | 1 | 2234 | 69 | |
| JQ907392.1 | - | 1 | 2215 | 69 | ||
More Information
Set Membership: |
|

MPEG1
Genomic Map