Assay Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315601.1 | NP_001302530.1 | 1 | 22 | 61 | |
| GenBank mRNA | JQ776642.1 | - | 1 | 23 | 61 | |
| JX566879.1 | - | 1 | 22 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315601.1 transcripts.
Set Membership: |
|

FFAR3
Genomic Map