Assay Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315676.1 | NP_001302605.1 | 3 - 4 | 296 | 68 | |
| GenBank mRNA | JQ740150.1 | - | 3 - 4 | 296 | 68 | |
More Information
Set Membership: |
|

ST8SIA2
Genomic Map