Assay Details
| Gene Symbol: NMUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315714.1 | NP_001302643.1 | 4 - 5 | 1282 | 58 | |
| XM_021074617.1 | XP_020930276.1 | 1234 | 58 | |||
| GenBank mRNA | KJ622336.1 | - | 4 - 5 | 1282 | 58 | |
More Information
Set Membership: |
|

NMUR1
Genomic Map