Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315720.1 | NP_001302649.1 | 6 - 7 | 640 | 59 | |
| GenBank mRNA | HQ397678.1 | - | 6 - 7 | 640 | 59 | |
More Information
Set Membership: |
|

ANKRD23
Genomic Map