Assay Details
| Gene Symbol: MTPAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317082.1 | NP_001304011.1 | 5 - 6 | 998 | 83 | |
| GenBank mRNA | AK234687.1 | - | 5 - 6 | 1002 | 83 | |
| GU345804.1 | - | 5 - 6 | 998 | 83 | ||
More Information
Set Membership: |
|

MTPAP
Genomic Map