Assay Details
| Gene Symbol: CD2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317083.1 | NP_001304012.1 | 4 - 5 | 546 | 73 | |
| XM_021098031.1 | XP_020953690.1 | 1151 | 73 | |||
| XM_021098032.1 | XP_020953691.1 | 1155 | 73 | |||
| GenBank mRNA | KJ410019.1 | - | 4 - 5 | 546 | 73 | |
More Information
Set Membership: |
|

CD2AP
Genomic Map