Assay Details
| Gene Symbol: UBE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257356.1 | NP_001244285.1 | 6 | 1221 | 91 | |
| XM_021078473.1 | XP_020934132.1 | 1182 | 91 | |||
| GenBank mRNA | AK400927.1 | - | 6 | 1257 | 91 | |
| JF272544.1 | - | 6 | 1221 | 91 | ||
More Information
Set Membership: |
|

UBE2B
Genomic Map