Assay Details
| Gene Symbol: TLR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113039.2 | NP_001106510.2 | 3 | 3190 | 87 | |
| NM_001293316.1 | NP_001280245.1 | 2 | 3023 | 87 | ||
| NM_001293317.1 | NP_001280246.1 | 3 | 1402 | 87 | ||
| NR_120680.1 | 3 | 1398 | 87 | |||
| GenBank mRNA | AB078418.1 | - | 3 | 2969 | 87 | |
| AK230475.1 | - | 3 | 1244 | 87 | ||
| AK230585.1 | - | 3 | 3039 | 87 | ||
| AK230797.1 | - | 1 | 1287 | 87 | ||
| AK343907.1 | - | 2 | 2881 | 87 | ||
| AK389696.1 | - | 3 | 1371 | 87 | ||
| AK396573.1 | - | 3 | 3190 | 87 | ||
| AK399479.1 | - | 1 | 2195 | 87 | ||
More Information
Set Membership: |
|

TLR4
Genomic Map