Assay Details
| Gene Symbol: ATG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190283.1 | NP_001177212.1 | 7 - 8 | 548 | 65 | |
| XM_021074594.1 | XP_020930253.1 | 509 | 65 | |||
| GenBank mRNA | AK395096.1 | - | 7 - 8 | 548 | 65 | |
| HM046508.1 | - | 7 - 8 | 548 | 65 | ||
More Information
Set Membership: |
|

ATG4B
Genomic Map