Assay Details
| Gene Symbol: SESN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356238.4 | XP_003356286.1 | 1538 | 55 | ||
| XM_021095665.1 | XP_020951324.1 | 3759 | 55 | |||
| XM_021095666.1 | XP_020951325.1 | 3610 | 55 | |||
More Information
Set Membership: |
|

SESN2
Genomic Map