Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483444.3 | XP_003483492.1 | 1443 | 94 | ||
| XM_013982988.2 | XP_013838442.1 | 450 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483444.3 | XP_003483492.1 | 1443 | 94 | ||
| XM_013982988.2 | XP_013838442.1 | 450 | 94 | |||
Set Membership: |
|