Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655354.3 | XP_005655411.1 | 148 | 99 | ||
| XM_021089065.1 | XP_020944724.1 | 148 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655354.3 | XP_005655411.1 | 148 | 99 | ||
| XM_021089065.1 | XP_020944724.1 | 148 | 99 | |||