Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655354.3 | XP_005655411.1 | 1173 | 120 | ||
| XM_021089063.1 | XP_020944722.1 | 1248 | 120 | |||
| XM_021089064.1 | XP_020944723.1 | 1248 | 120 | |||
| XM_021089065.1 | XP_020944724.1 | 1173 | 120 | |||
More Information
Set Membership: |
|

ERICH5
Genomic Map