Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662778.3 | XP_005662835.1 | 417 | 98 | ||
| XM_005662781.3 | XP_005662838.1 | 222 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COLEC11| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662778.3 | XP_005662835.1 | 417 | 98 | ||
| XM_005662781.3 | XP_005662838.1 | 222 | 98 | |||
Set Membership: |
|