Assay Details
| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984592.2 | XP_013840046.1 | 318 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013984592.2 transcripts.
Set Membership: |
|

NEU2
Genomic Map