Assay Details
| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984592.2 | XP_013840046.1 | 526 | 102 | ||
| XM_021074906.1 | XP_020930565.1 | 543 | 102 | |||
| XM_021074907.1 | XP_020930566.1 | 568 | 102 | |||
More Information
Set Membership: |
|

NEU2
Genomic Map