Assay Details
| Gene Symbol: C16H5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984941.2 | XP_013840395.1 | 808 | 79 | ||
| XM_013984942.2 | XP_013840396.1 | 804 | 79 | |||
More Information
Set Membership: |
|

C16H5orf38
Genomic Map