Assay Details
| Gene Symbol: C16H5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984941.2 | XP_013840395.1 | 919 | 63 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H5orf38| Gene Symbol: C16H5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984941.2 | XP_013840395.1 | 919 | 63 | ||