Assay Details
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996393.2 | XP_013851847.2 | 524 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996393.2 | XP_013851847.2 | 524 | 98 | ||