Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927997.5 | XP_001928032.1 | 2326 | 71 | ||
| XM_001928002.4 | XP_001928037.1 | 2268 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927997.5 | XP_001928032.1 | 2326 | 71 | ||
| XM_001928002.4 | XP_001928037.1 | 2268 | 71 | |||
Set Membership: |
|