Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928721.5 | XP_001928756.1 | 976 | 69 | ||
| XM_005671297.3 | XP_005671354.1 | 978 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928721.5 | XP_001928756.1 | 976 | 69 | ||
| XM_005671297.3 | XP_005671354.1 | 978 | 69 | |||
Set Membership: |
|