Assay Details
| Gene Symbol: LOC110255463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063176.1 | XP_020918835.1 | 1079 | 95 | ||
| XM_021063177.1 | XP_020918836.1 | 1014 | 95 | |||
| XM_021063178.1 | XP_020918837.1 | 1029 | 95 | |||
| XM_021063179.1 | XP_020918838.1 | 1738 | 95 | |||
| XM_021063180.1 | XP_020918839.1 | 1644 | 95 | |||
| XM_021063181.1 | XP_020918840.1 | 921 | 95 | |||
More Information
Set Membership: |
|

LOC110255463
Genomic Map