Assay Details
| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063516.1 | XP_020919175.1 | 918 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC71L| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063516.1 | XP_020919175.1 | 918 | 108 | ||
Set Membership: |
|