Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067089.1 | XP_020922748.1 | 248 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021067089.1 transcripts.
Set Membership: |
|

PRR15L
Genomic Map