Assay Details
| Gene Symbol: TPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122941.3 | XP_003122989.1 | 1089 | 80 | ||
| XM_005661097.2 | XP_005661154.1 | 1238 | 80 | |||
| XM_021083266.1 | XP_020938925.1 | 963 | 80 | |||
More Information
Set Membership: |
|

TPH1
Genomic Map