Assay Details
| Gene Symbol: PTGER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123373.4 | XP_003123421.3 | 2356 | 71 | ||
| XM_021083693.1 | XP_020939352.1 | 1333 | 71 | |||
More Information
Set Membership: |
|

PTGER1
Genomic Map