Assay Details
| Gene Symbol: LOC100153087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080220.1 | XP_020935879.1 | 84 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021080220.1 transcripts.
Set Membership: |
|

LOC100153087
Genomic Map