Assay Details
| Gene Symbol: LOC102160746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082418.1 | XP_020938077.1 | 687 | 58 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102160746| Gene Symbol: LOC102160746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082418.1 | XP_020938077.1 | 687 | 58 | ||