Assay Details
| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093735.1 | XP_020949394.1 | 967 | 68 | ||
| XM_021093736.1 | XP_020949395.1 | 870 | 68 | |||
| XM_021093737.1 | XP_020949396.1 | 980 | 68 | |||
More Information
Set Membership: |
|

SLC38A8
Genomic Map