Assay Details
| Gene Symbol: LOC102164667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096470.1 | XP_020952129.1 | 2532 | 100 | ||
| XM_021096471.1 | XP_020952130.1 | 2475 | 100 | |||
| XM_021096473.1 | XP_020952132.1 | 2552 | 100 | |||
More Information
Set Membership: |
|

LOC102164667
Genomic Map