Assay Details
| Gene Symbol: LOC102164667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096470.1 | XP_020952129.1 | 650 | 84 | ||
| XM_021096471.1 | XP_020952130.1 | 650 | 84 | |||
| XM_021096472.1 | XP_020952131.1 | 650 | 84 | |||
| XM_021096473.1 | XP_020952132.1 | 650 | 84 | |||
| XM_021096474.1 | XP_020952133.1 | 650 | 84 | |||
| XM_021096476.1 | XP_020952135.1 | 650 | 84 | |||
| XM_021096477.1 | XP_020952136.1 | 650 | 84 | |||
More Information
Set Membership: |
|

LOC102164667
Genomic Map