Assay Details
| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021102698.1 | XP_020958357.1 | 483 | 79 | ||
| XM_021102699.1 | XP_020958358.1 | 478 | 79 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021102698.1 | XP_020958357.1 | 483 | 79 | ||
| XM_021102699.1 | XP_020958358.1 | 478 | 79 | |||