Assay Details
| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129941.4 | XP_003129989.1 | 1275 | 62 | ||
| XM_013979475.2 | XP_013834929.1 | 1502 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129941.4 | XP_003129989.1 | 1275 | 62 | ||
| XM_013979475.2 | XP_013834929.1 | 1502 | 62 | |||
Set Membership: |
|