Assay Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133731.3 | XP_003133779.1 | 1156 | 63 | ||
| XM_021074830.1 | XP_020930489.1 | 1355 | 63 | |||
| XM_021074831.1 | XP_020930490.1 | 1352 | 63 | |||
More Information
Set Membership: |
|

CHRNG
Genomic Map