Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135329.5 | XP_003135377.2 | 1055 | 75 | ||
| XM_021079889.1 | XP_020935548.1 | 1168 | 75 | |||
More Information
Set Membership: |
|

AKAP14
Genomic Map