Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998442.2 | XP_013853896.1 | 3207 | 97 | ||
| XM_013998444.2 | XP_013853898.1 | 598 | 97 | |||
| XM_013998445.2 | XP_013853899.1 | 364 | 97 | |||
More Information
Set Membership: |
|

ZNF529
Genomic Map